Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosflow Perm Buffer II, BD

Catalog No. BDB558052 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558052 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558052 Supplier BD Biosciences Supplier No. 558052
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For the permeabilization of fixed Peripheral Blood Mononuclear Cells (PBMCs) or lysed whole blood.

  • Designed for optimal fluorescent staining in flow cytometry
  • Can be used with antibodies for phospho-protein analysis
  • In many cases, may be used simultaneously with phospho-specific antibodies and the staining of cell surface markers (e.g CD markers)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Format Liquid
Quantity 125 mL
Validated Application RUO
Product Type Staining Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.