Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Plated Media M-ENDO Agar

Catalog No. B98291 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
20/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B98291 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B98291 Supplier BD Supplier No. 298291
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Slightly selective and differential medium for isolation and differentiation of Enterobacteriaceae

  • Used in enumeration of coliforms in water by the membrane filter technique
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Purple
Content And Storage <30°C
Form Solid
Format Prepared Plate
Manufacturing Origin United States
Media Type m Endo Agar
Product Type Prepared Microbiology Plate
Packaging Type Plate
Quantity 20/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.