Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Plated Media Trypticase™ Soy Broth

Catalog No. B257202 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10,000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B257202 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B257202 Supplier BD Supplier No. 257202
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Amber
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy Broth
Packaging Type Bag
Quantity 10,000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.