Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Trypticase™ Plated Media: Soy Broth

Catalog No. L4399749 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Certifications/Compliance:
ISO/DIS, 11014
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
L4399749 ISO/DIS, 11014
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L4399749 Supplier BD Supplier No. 299749
Add to Cart
Add to Cart

Used to differentiate Enterococcus spp. from the Streptococcus bovis group of streptococci

  • General purpose medium for cultivation of fastidious and nonfastidious microorganisms
  • With addition of 6.5% sodium chloride, differentiation between salt-tolerant and salt-intolerant species is achieved
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Beige
Content And Storage 2 to 25°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy broth
No. per Pack 100
Packaging Type Tube Size A
Quantity 20 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.