Learn More
Plus Permeabilization Buffer, BD
Catalog No. BDB561651 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 mL
Catalog No. Quantity
BDB561651 10 mL
1 options
Catalog No. BDB561651 Supplier BD Biosciences Supplier No. 561651
Add to Cart
Add to Cart
Specially formulated for the immunofluorescent staining of incorporated BrdU for flow cytometric analysis
  • Used as a staining enhancer and secondary permeabilization reagent
  • Should be used with fixed cell samples only
Flow Cytometry, Intracellular staining
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Permeabilization Buffer
For Use With (Application) Immunofluorescent staining of incorporated BrdU for flow cytometric analysis, staining enhancer and secondary permeabilization reagent, intracellular staining
Quantity 10 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov