Learn More
Description
Includes
Kit includes one 2.5mL vial of S. pneumoniae antibody-coated latex bead suspension, one vial of lyophilized polyvalent positive control (reconstitutes to 5mL), one 2.5mL vial with dropper, latex negative control, 50 stirring sticks and one glass slide.
- Test results within three minutes
- CLIA Complexity: High
- Qualitatively detects capsular antigens of S. pneumoniae directly from isolated colonies or pure cultures from liquid nutrient broth
- Visible agglutination occurs when the S. pneumoniae capsular antigen reacts with the antibody-coated latex beads
- Sensitivity: 98.4% with colonies isolated on agar
- Specificity: 93.0% with colonies isolated on agar
Specifications
Specifications
| Clia Complexity | High |
| Detectable Analytes | S. pneumoniae Capsular Antigen |
| Sample Type | Isolated colonies or pure cultures from liquid nutrient broth |
| Type | S. pneumoniae Latex Test Kit |
| Includes | One 2.5mL vial of Antibody-coated Latex bead suspension, One 0.5mL vial of lyophilized positive control, One 2.5mL vial of negative control, 50 Stirring sticks and One glass slide |
| Quantity | 50 Tests |
| Test Time | 3 min. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.