Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Prepared Bottled Media: Fluid Thioglycollate with 1% Tween

Catalog No. 22349722 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-349-722 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-349-722 Supplier BD Supplier No. 257365
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples

Ready for immediate use

TRUSTED_SUSTAINABILITY

Specifications

Appearance Light Beige to Light Amber, Clear or Slightly Hazy, Free of Precipitate or Excessive Debris
Broth Type Fluid Thioglycollate Medium
Cap Type Screw Cap
Color Amber
Content And Storage 2°C to 30°C
Additive Polysorbate 80
Detects Microorganisms
Form Liquid
Format Bottle
Manufacturing Origin United States
Media Type Fluid Thioglycollate with 1% Tween
Preparation Method Ready-to-Use
Product Type Liquid Medium
Target Organism Class Bacteria
Test Type Microbial Detection
Fill Volume 500 mL
For Use With (Application) Detection of microorganisms from pharmaceutical, food, environmental or Water testing samples
Includes Septum
Incubation Conditions 30°C to 35°C
Incubation Time 18 to 72 hr.
Packaging Type Bottle
Quantity 500 mL
Specimen Type Pharmaceutical, Food, Environmental or Water Testing Samples
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.