Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Prepared Bottled Media: Sabouraud Dextrose Broth

Catalog No. 22349973 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Sabouraud Dextrose Broth; 100mL; 10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-349-973 Sabouraud Dextrose Broth; 100mL; 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-349-973 Supplier BD Supplier No. 215193
Add to Cart
Add to Cart
This item is not returnable. View return policy

For the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples

Ready for immediate use

TRUSTED_SUSTAINABILITY

Specifications

Appearance Amber
Broth Type Sabouraud Dextrose Broth
Cap Type Screw Cap
Color Amber
Content And Storage 2°C to 8°C
Country of Origin United States
Detects Microorganisms, Candida Albicans
Form Liquid
Format Bottle
Manufacturing Origin United States
Media Type Sabouraud Dextrose Broth
Preparation Method Ready-to-Use
Product Type Broth
Sterility Sterile
Target Organism Class Molds
Test Type Microbial Detection
Fill Volume 100 mL
Final Product Type Liquid Medium
For Use With (Application) Detection of microorganisms from pharmaceutical, food, environmental or Water testing samples
Packaging Type Bottle
Quantity 10/Pk.
Specimen Type Pharmaceutical, Food, Environmental, Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.