Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Prepared Bottled Media: Trypticase™ Soy Broth (Double Wrapped) Infusion Bottle

Catalog No. 22349731 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-349-731 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-349-731 Supplier BD Supplier No. 257265
Add to Cart
Add to Cart

For the detection of microorganisms from pharmaceutical, food, environmental, or water testing samples

Ready for immediate use

TRUSTED_SUSTAINABILITY

Specifications

Appearance Amber
Broth Type Tryptic Soy Broth
Cap Type Crimp
Color Amber
Content And Storage 5°C to 25°C
Country of Origin United States
Detects Microorganisms
Form Liquid
Format Bottle
Manufacturing Origin United States
Media Type Trypticase Soy Broth
Preparation Method Ready-to-Use
Product Type Broth
Sterility Sterile
Test Type Microbial Detection
Fill Volume 100 mL
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Glucose (Dextrose) 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L
For Use With (Application) Detection of micororganisms from pharmaceutical, food, environmental or Water testing samples
Includes Removable Septum Stopper
Packaging Type Bottle
Quantity 10/Pk.
Specimen Type Pharmaceutical Samples, Food Samples, Environmental Samples, Water Samples
Protein Source Casein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.