Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Prepared Bottled Media: Trypticase™ Soy Broth, Double Wrapped, Twist-off, EP/USP Wide Mouth Jar

Catalog No. 22349991 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22349991 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22349991 Supplier BD Supplier No. 257294
Add to Cart
Add to Cart

For sterility testing in sterile filling rooms

Ready for immediate use

TRUSTED_SUSTAINABILITY

Specifications

Appearance Beige
Broth Type Tryptic Soy Broth
Color Beige
Country of Origin United States
Detects Microorganisms, Bacteria, Yeast, Fungi
Form Liquid
Format Wide Mouth Jar
Manufacturing Origin United States
Media Type Trypticase Soy Broth (double wrapped), twist-off, EP/USP
Product Type Culture Medium
Sterility Sterile
Test Type Sterility Testing
Fill Volume 100 mL
Final Product Type Microbial Growth
Formulation Pancreatic Digest of Casein-17.0 gm, Sodium Chloride-5.0 gm, Papaic Digest of Soybean Meal-3.0 gm, Dextrose-2.5 gm, Dipotassium Phosphate-2.5 gm
For Use With (Application) Detection of microorganisms from pharmaceutical, food, environmental or Water testing samples
Incubation Time 18 to 24 hr.
Packaging Type Bottle
Product Line BD BBL
Quantity 100 mL
Specimen Type Pharmaceutical, Food, Environmental, Water
Protein Source Casein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.