Learn More
Description
Ready for immediate use
Specifications
Specifications
| Color | Amber |
| Content And Storage | 5°C to 25°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Trypticase Soy Broth |
| Fill Volume | 500 mL |
| Formulation | Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Glucose (Dextrose) 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L |
| For Use With (Application) | Detection of micororganisms from pharmaceutical, food, environmental or Water testing samples |
| Packaging Type | Bottle |
| Quantity | 500 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.