Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Prepared Media R2A 100 mm Agar Plate

Catalog No. b292341 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Color:
Light Amber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B292341 Light Amber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B292341 Supplier BD Supplier No. 292341
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Yeast extract 0.5g/L; Proteose peptone No. 3 0.5g/L; Casamino Acids 0.5g/L; Dextrose 0.5g/L; Soluble starch 0.5g/L; Sodium pyruvate 0.3g/L; Dipotassium phosphate 0.3g/L; Magnesium sulfate 0.05g/L; Agar 15.0g/L

Improved recovery of stressed and chlorine-tolerant organisms

  • Format: Agar Plates
  • Appearance: Light amber, slightly opalescent with a slight precipitate
  • pH at 25°C: 7.2 ± 0.2
  • Sterile

For plate counts of heterotrophic organisms in treated potable water

TRUSTED_SUSTAINABILITY

Specifications

Color Light Amber
Format Agar Plates
Manufacturing Origin United States
Media Type R2A Agar
Sterility Sterile
For Use With (Application) Treating potable Water against compromised bacteria
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.