Learn More
BD ProbeTec™ Qx Collection Kit for Endocervical and Lesion Specimens
Shop All Waters Corporation ProductsDescription
Includes
Each kit includes one sterile polyester fiber-tipped cleaning swab with a white shaft, one sterile polyurethane foam-tipped collection swab with a pink shaft, one tube of Qx Swab Diluent
BD ProbeTec™ Qx Collection Kit
- For collection and transportation of endocervical or lesion specimens for testing on the BD Viper™ System in extracted mode
- Endocervical specimens are to be used with the BD ProbeTec CT/GC Qx Amplified DNA Assays
- External anogenital lesion specimens are to be used with the BD ProbeTec HSV1&2 Qx Amplified DNA Assays
- A polyester fiber-tipped cleaning swab is provided to clean the cervical os of blood and mucus prior to endocervical specimen collection
- A polyurethane foam-tipped swab is used to collect the specimen for testing
Specifications
Specifications
| Product Type | QX Collection Kit |
| Includes | Sterile Polyester fiber-tipped cleaning swab with a white shaft, sterile Polyurethane foam-tipped collection swab with a pink shaft, one tube of Qx swab diluent |
| For Use With (Application) | For endocervical CT/GC or external anogenital lesion HSV1 and 2 specimens collection |
| For Use With (Equipment) | BD viper systems |
| Quantity | Each |