Learn More
BD ProbeTec™ Qx CT/GC Urethral Specimen Collection Kits
Catalog No. 22370172 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
Pack of 100
Catalog No. Quantity
22-370-172 Pack of 100
1 options
Catalog No. 22-370-172 Supplier BD Supplier No. 441358
This product may not be ordered online. Please contact your sales representative for further ordering instructions.
Includes
Pack of 100
For collection and transportion of male urethral specimens to the laboratory for CT/GC testing
Male Urethral Specimen Collection Kit
  • Used to collect and transport male patient urethral specimens to the laboratory for testing
  • The rayon-tipped collection swab consists of a stainless steel wire in a blue-colored plastic shaft that is scored for breaking
  • For the BD ProbeTec™ Chlamydia trachomatis/Neisseria gonorrhoeae (CT/GC) Qx Amplified DNA Assays and BD Viper™ System in extracted mode
TRUSTED_SUSTAINABILITY
Specifications
Product Type Specimen Collection Kit
For Use With (Application) Used to collect and transport male patient urethral specimens to the laboratory for testing
Includes Rayon-tipped collection swab consists of a Stainless Steel wire, Blue Plastic shaft
Quantity Pack of 100