Learn More
BD Propidium Iodide Staining Solution
Catalog No. BDB556463 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
BDB556463 2 mL
1 options
Catalog No. BDB556463 Supplier BD Supplier No. 556463
Add to Cart
Add to Cart
Designed for use in two-color Annexin V flow cytometric assays.
  • Detected in the orange range of the spectrum using a 562-588nm band pass filter
  • Used to assess plasma membrane (PM) integrity in Annexin V apoptosis assays
Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 mL
Product Type Pharmingen Propidium Iodine Staining Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov