Learn More
BD Cell Analysis 3P BD QUANTIBRITETM PE BEADS
Catalog No. 50620179
Encompass
QuantiBRITE PE beads are used with PE-labeled monoclonal antibodies to estimate antibodies bound per cell (ABC) by flow cytometry.
Catalog No. 50-620-179 Supplier BD Cell Analysis 3P Supplier No. 340495
QuantiBRITE PE beads are used with PE-labeled monoclonal antibodies to estimate antibodies bound per cell (ABC) by flow cytometry.