Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Rat IL-4 ELISA Set

Catalog No. BDB555198 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555198 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555198 Supplier BD Supplier No. 555198
Add to Cart
Add to Cart

For rat interleukin-4 (IL-4)

The OptEIA™ set contains components necessary to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant rat IL-4 in serum, plasma, and cell culture supernatants.

  • Capture antibody: anti-rat IL-4 monoclonal antibody
  • Detection antibody: biotinylated anti-rat IL-4 monoclonal antibody
  • Enzyme reagent: Streptavidin-horseradish peroxidase conjugate
  • Standards: lyophilized recombinant rat IL-4
  • Standard range: 1.6 - 100pg/mL

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.6 to 100pg/mL
Product Type Rat IL-4 ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Rat IL-4 in Serum, Plasma and cell culture supernatants
Kit Contents anti-Rat IL-4 monoclonal (capture) antibody, Biotinylated anti-Rat IL-4 monoclonal (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Rat IL-4 (standard), Streptavidin-HRP Conjugate
Quantity 20 Plates
Storage Requirements 2°C to 8°C

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.