Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Rat TNF ELISA Kit

Catalog No. BDB560479 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
2 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560479 2 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560479 Supplier BD Supplier No. 560479
Add to Cart
Add to Cart

For the quantitative determination of TNF in rat serum, plasma, and cell culture supernatant

  • Solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay)
  • Utilizes a monoclonal antibody specific for rat TNF coated on a 96-well plate

TRUSTED_SUSTAINABILITY

Specifications

Antigen TNF
Cross Reactivity Rat
Includes Stop solution, standard/Sample diluent, TMB one-step Substrate reagent, PerCP-Cy™5.5 Mouse Anti-Human CD13, Enzyme diluent, rat TNF Antibody coated plate, rat TNF standard, Enzyme concentrate Streptavidin HRP, wash concentrate (20X), det
Quantity 2 Plates
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Quantitative determination of TNF, ELISA
Product Type Rat TNF ELISA kit

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.