Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Rat TNF ELISA Set

Catalog No. BDB558535 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558535 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558535 Supplier BD Supplier No. 558535
Add to Cart
Add to Cart

Includes

Capture Antibody: Anti-rat TNF-α, Detection Antibody: Biotinylated anti-rat TNF-α, Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP) (250X), Standard: Recombinant rat TNF-α (lyophilized)

Design an immunoassay with different sensitivity and dynamic range

  • Flexible assay
  • Calibrated against purified Baculovirus-expressed recombinant rat TNF

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Concentration 31.3 to 2000pg/mL
Product Type Rat TNF ELISA set
Sufficient For 20 96-well Plates
For Use With (Application) To develop ELISA for natural or recombinant Rat TNF in Serum, Plasma and cell culture supernatants
Kit Contents Biotin anti-Rat TNF (250X) (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Rat TNF (standard), Purified anti-Rat TNF (250X) (capture) antibody, Streptavidin-HRP Conjugate (250X)
Quantity 20 Plates
Storage Requirements 2°C to 8°C

Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.