Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Nitrate B Reagent Dropper

Catalog No. 1491062 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
50 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-62 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-910-62 Supplier BD Supplier No. 261198
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Identify microorganisms without mixing or measuring reagents

To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.

  • Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
  • Strict quality controls assure uniformity and accuracy

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 Pack
Certifications/Compliance CE
Format Dropper
For Use With (Application) For use in determining the ability of bacteria to reduce nitrate to nitrite or free nitrogen gas
Storage Requirements 15° to 30°C
Stain Type Nitrate Reagent B
Volume (Metric) 0.5 mL
Product Type Reagent/Stain Dropper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.