Learn More
Description
To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.
- Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
- Strict quality controls assure uniformity and accuracy
Specifications
Specifications
| Quantity | 50 Pack |
| Certifications/Compliance | CE |
| Format | Dropper |
| For Use With (Application) | Used for presumptive differentiation of pneumococci based on bile solubility |
| Storage Requirements | 15° to 30°C |
| Stain Type | Desoxycholate |
| Volume (Metric) | 0.5 mL |
| Product Type | Reagent/Stain Dropper |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.