Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Erythropoietin (EPO) Protein

Catalog No. BDB554597 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554597 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554597 Supplier BD Supplier No. 554597
Add to Cart
Add to Cart

Determined by SDS-PAGE and an absorbance assay based on Beers-Lambert law

30 kD heavily glycosylated protein containing 166 amino acids

  • Reactivity: Mouse
  • Storage Buffer: Frozen aqueous buffered solution containing BSA and glycerol
  • Measured using TF-1 indicator cells
  • Specific Activity: 0.1 - 1.0 × 108 Units/mg (Unit is defined as the amount of material required to stimulate a half-maximal response at cytokine saturation)
  • ED50: 0.1 to 1.0 ng/ml; Observed linear dose response range: >100 fold

Bioassay

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.1mg/mL
For Use With (Application) Bioassay
Formulation Frozen aqueous Buffered solution containing BSA and Glycerol
Molecular Weight (g/mol) 30
Name Mouse Erythropoietin Recombinant Protein
Quantity 5 μg
Storage Requirements -80°C
Cross Reactivity Mouse
Recombinant Recombinant
Form Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.