Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Human β2 Microglobulin

Catalog No. BDB551089 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB551089 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB551089 Supplier BD Supplier No. 551089
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant protein expressed in insect cells and purified by a combination of ion-exchange and gel-filtration methods

  • Storage buffer: Aqueous buffered solution containing ≤0.09% sodium azide
  • Maximal activity range: 50-200ng/mL

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Type Beta-2 Microglobulin
Quantity 0.1 mg

Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.