Learn More
Description
Human Eotaxin:
- 74-amino acid protein
- Molecular weight of 8.4kDa
- Belongs to C-C chemokine family
- Research use only
- Acts as a potent and selective chemoattractant for eosinophils, but not monocytes, lymphocytes or neutrophils
Specifications
Specifications
| For Use With (Application) | ELISA, Functional assay |
| Product Type | Human Eotaxin |
| Quantity | 5 μg |
| Content And Storage | -80°C |
| Format | Frozen |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.