Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Human GM-CSF

Catalog No. BDB550068 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550068 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550068 Supplier BD Supplier No. 550068
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stimulates precursor cells of granulocytes, macrophages, and eosinophils

Quantitative standard for measuring human GM-CSF protein levels in a GM-CSF specific sandwich ELISA with the purified BVD2-23B6 antibody as capture antibody and the biotinylated BVD2-21C11 as detection antibody

  • Reactivity: Human
  • Storage buffer: Frozen aqueous buffered solution containing BSA
  • Biological Activity: Proliferation measured using TF-1 indicator cells
  • Specific Activity: 0.1- 1.0 x 109 Unit/mg
  • ED50: 10 - 90pg/ml

Bioassay, Enzyme-linked immunosorbent assay standard, Flow cytometry block

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 80°C
Quantity 10 μg
Source Human

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.