Learn More
Description
- Supplied as a frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution containing 1mg/mL Biotechnology grade, low endotoxin bovine serum albumin, with no preservatives
Enzyme Linked Immunosorbent Assay, BioAssay
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard |
| Name | IL-12 (p70) Recombinant Protein |
| Quantity | 5 μg |
| Storage Requirements | -80°C |
| Recombinant | Recombinant |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.