Learn More
Description
- Comes as a frozen aqueous buffered solution containing glycerol, 2.0mg/mL Biotechnology grade, low endotoxin bovine serum albumin, and with no preservatives
- Can be used as a blocking control to demonstrate specificity of IL-3 staining by the directly conjugated-BVD3 1F9 antibody
Enzyme Linked Immunosorbent Assay, BioAssay
Specifications
Specifications
| Concentration | 200μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard, Intracellular Block/Flow Cytometry |
| Name | Human IL-3 Recombinant Protein |
| Quantity | 10 μg |
| Storage Requirements | -80°C |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.