Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Human Interleukin-1β (IL-1β)

Catalog No. BDB554602 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554602 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554602 Supplier BD Supplier No. 554602
Add to Cart
Add to Cart

Determined by SDS-PAGE analysis, and an absorbance assay based on the Beers-Lambert law

Lyophilized from a solution comprised of 0.22 ++m sterile-filtered aqueous buffered solution with no preservatives

  • Reactivity: Human
  • Storage Buffer: Lyophilized in an aqueous buffered solution containing BSA
  • Biological Activity: The activity range is measured in a proliferation assay using TF-1 indicator cells
  • Specific Activity Range: 1.0-4.0x108 Units/mg
  • ED50: 25 to 100pg/ml
  • Observed dose response: 100 fold

Bioassay

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioassay
Formulation Lyophilized in an aqueous Buffered solution containing BSA
Name Human IL-1beta Recombinant Protein
Purification Method SDS-PAGE
Quantity 5 μg
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant
Form Lyophilized

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.