Learn More
Description
Frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution, pH 7.2, and containing biotechnology grade, low endotoxin bovine serum albumin, with no preservatives
- Reactivity: Human
- Storage Buffer: Frozen aqueous buffered solution containing BSA
- Biological Activity: The activity range is measured in a proliferation assay using TF-1 indicator cells
- Specific Activity: 0.4 - 5.0 × 108 Units/mg
- ED50: 20 - 250pg/ml
- Observed dose response relationship: 100 fold
Enzyme-linked immunosorbent assay, bioassay
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard |
| Formulation | Frozen aqueous Buffered solution containing BSA |
| Molecular Weight (g/mol) | 15 |
| Name | Human IL-4 Recombinant Protein |
| pH Range | 7.2 |
| Quantity | 5 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Show More |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.