Learn More
Description
Frozen liquid comprised of 0.22 ++m sterile-filtered aqueous buffered solution (pH 7.2) and containing 2 mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives
- Reactivity: Human
- Storage Buffer: Frozen aqueous buffered solution containing BSA
Enzyme-linked immunosorbent assay standard
Specifications
Specifications
| For Use With (Application) | Bioassay, ELISA Standard |
| Formulation | Frozen aqueous Buffered solution containing BSA |
| Name | Human MCP-1 Recombinant Protein |
| Quantity | 10 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
| Form | Solution |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.