Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Human RANTES

Catalog No. BDB557910 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557910 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557910 Supplier BD Supplier No. 557910

A potent chemoattractant for monocytes, T lymphocytes, basophils, eosinophils, NK cells and dendritic cells

  • Belongs to CC chemokine family
  • Acts as a potent chemoattractant for monocytes, T lymphocytes, basophils, eosinophils, NK cells and dendritic cells

Bioassay, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
For Use With (Application) ELISA, Functional assay
Product Type Human RANTES
Quantity 10 μg
Storage Buffer Frozen Aqueous Buffered solution with BSA
Content And Storage -80°C
Reactivity Human
Format Frozen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.