Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Human Tumor Necrosis Factor(TNF-α)

Catalog No. bdb554618 Shop All BD Biosciences Products
Change view
Click to view available options
Concentration:
200μg/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
BDB554618 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554618 Supplier BD Supplier No. 554618
Only null left
Add to Cart
Add to Cart

Pure by SDS-PAGE analysis, and an absorbance assay based on the Beers-Lambert law

Frozen liquid comprised of 0.22μm sterile-filtered PBS, pH 7.2, and containing 4mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no preservatives

  • Reactivity: Human
  • Storage Buffer: Frozen aqueous buffered solution containing BSA
  • Biological Activity Range: Measured in a cytolysis assay using L929 indicator cells
  • Specific activity: 0.05 - 2.0 X 109 Units/mg
  • ED50: 5 TO 200pg/ml
  • Observed Dose response: 100 fold

Bioassay. enzyme-linked immunosorbent assay standard

TRUSTED_SUSTAINABILITY

Specifications

Concentration 200μg/mL
For Use With (Application) Bioassay, ELISA Standard, ELISA
Formulation Frozen aqueous Buffered solution containing BSA
Name Human TNF-a Recombinant Protein
Quantity 10μg
Storage Requirements -80°C
Cross Reactivity Human
Recombinant Recombinant

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.