Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Mouse GM-CSF

Catalog No. BDB554586 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554586 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554586 Supplier BD Supplier No. 554586
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Has potent stimulatory effects on growth and differentiation of bone marrow progenitor cells

  • Produced by activated T cells, macrophages, vascular endothelial cells and fibroblasts
  • Contains 124 amino acid residues
  • Helps in generating granulocytes, monocytes/macrophages, and megakaryocytes
  • Act on mature leukocytes to promote inflammatory responses in peripheral tissues
  • Appears as 15.5 - 19kD band in reducing and non-reducing SDS-PAGE analysis
  • Storage buffer: frozen aqueous buffered solution containing BSA

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C
Quantity 10 μg
Source Mouse
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.