Learn More
Description
- Purified by immunoaffinity chromatography
Enzyme Linked Immunosorbent Assay, BioAssay
Specifications
Specifications
| Concentration | 200μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard |
| Name | Mouse IFN-gamma Recombinant Protein |
| Quantity | 10 μg |
| Storage Requirements | -80°C |
| Recombinant | Recombinant |
| Form | Liquid |
Safety and Handling
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.