Learn More
Description
Quantitative standard for measuring mouse MCP-1 protein levels in a sandwich ELISA with the purified 2H5 antibody as a capture antibody and the biotinylated 4E2/MCP antibody as the detection antibody
- Reactivity: Mouse
- ED50: 50 - 800ng/ml
Bioassay, Enzyme-linked immunosorbent assay standard
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | Bioassay, ELISA Standard |
| Name | Mouse MCP-1 Recombinant Protein |
| Quantity | 5 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Mouse |
| Recombinant | Recombinant |
Safety and Handling
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.