Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Mouse TNF

Catalog No. BDB554589 Shop All BD Biosciences Products
Change view
Click to view available options
Concentration:
200μg/mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
BDB554589 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554589 Supplier BD Supplier No. 554589
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent multifunctional cytokine containing 156 amino acid residues

  • Glycosylated protein
  • Exerts regulatory and cytotoxic effects on wide range of normal lymphoid and non-lymphoid cells and tumor cells
  • Storage buffer: frozen aqueous buffered solution containing BSA

Bioassay, ELISA standard, blocking

TRUSTED_SUSTAINABILITY

Specifications

Concentration 200μg/mL
For Use With (Application) Bioassay, Blocking Agent, ELISA Standard
Formulation Frozen aqueous Buffered solution containing BSA
Name TNF-alpha Recombinant Protein
Quantity 10μg
Storage Requirements -80°C
Cross Reactivity Mouse
Recombinant Recombinant

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.