Learn More
Description
- Molecular weight: 18.5kD protein
- Inhibits macrophage-mediated cytokine synthesis
- Suppresses delayed type hypersensitivity response
- Stimulate Th2 cell response
- Helps in elevated antibody production
- Storage buffer: frozen aqueous buffered solution containing BSA
ELISA standard, intracellular block/flow cytometry
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | ELISA Standard, Intracellular Block/Flow Cytometry |
| Formulation | Frozen aqueous Buffered solution containing BSA |
| Molecular Weight (g/mol) | 18.5 |
| Name | Rat IL-10 Recombinant Protein |
| Quantity | 5 μg |
| Storage Requirements | -80°C |
| Cross Reactivity | Rat |
| Recombinant | Recombinant |
| Form | Solution |
For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.