Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Rat Interleukin-4 (IL-4)

Catalog No. BDB555107 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μ g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555107 5 μ g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555107 Supplier BD Supplier No. 555107
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent lymphoid cell growth factor

  • Stimulate growth, differentiation, survivability, function of cells of hematopoietic and non-hematopoietic origin
  • Binds to specific high affinity receptors
  • Gene product is 17 -20 kD glycoprotein containing 123 amino acid residues
  • Storage buffer: frozen aqueous buffered solution containing BSA

ELISA standard, bioassay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C
Quantity 5 μ g
Source Rat
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.