Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recombinant Rat Monocyte chemoattractant protein-1 (MCP-1)

Catalog No. BDB555110 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555110 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555110 Supplier BD Supplier No. 555110
Add to Cart
Add to Cart

Highly glycosylated protein with chemoattractant activity for monocytes, lymphocytes and basophils

  • Member of C-C chemokine superfamily
  • Homolog of human MCP-1
  • Reactivity: rat
  • Size: 27 - 30kD
  • Expressed by variety of cell types including monocyte/macrophages, endothelial cells and mesangial cells
  • Supplied as frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution
  • Contains glycerol, 1.0mg/mL biotechnology grade, low endotoxin bovine serum albumin
  • Storage buffer: Frozen aqueous buffered solution containing BSA and glycerol

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.1 mg/mL
Type PharMingen™ Recombinant Rat MCP-1
For Use With (Application) ELISA standard, bioassay, intracellular block/flow cytometry
Includes Glycerol 1.0mg/mL biotechnology grade, low endotoxin bovine Serum Albumin
Quantity 5 μg
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.