Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Recomninant Rat GM-CSF

Catalog No. BDB555111 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 μ g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555111 5 μ g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555111 Supplier BD Supplier No. 555111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytokine having potent stimulatory effects on growth and differentiation of bone marrow progenitor cells

  • Produced by activated T cells, macrophages, vascular endothelial cells and fibroblast
  • Acts on mature leukocytes to promote inflammatory responses in peripheral tissues
  • Promotes inflammatory responses
  • Storage buffer: frozen aqueous buffered solution containing BSA and glycerol

ELISA standard, bioassay

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -80°C
Quantity 5 μ g
Source Rat
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.