Learn More
BD Cell Analysis 3P Reticulocyte Whole Blood Control, 200 tests
Catalog No. 50620459
Encompass
The BD Retic-Count control is a tri-level, assayed hematology control designed to document and monitor values obtained from manual and automated reticulocyte counting methods.
Catalog No. 50-620-459 Supplier BD Cell Analysis 3P Supplier No. 340999
The BD Retic-Count control is a tri-level, assayed hematology control designed to document and monitor values obtained from manual and automated reticulocyte counting methods.