Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Retrievagen Antigen Retrieval Systems

Catalog No. BDB550527 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
125 mL
250 mL
Reagent Type:
Retrievagen A
Retrievagen B
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Reagent Type
BDB550527 250 mL Retrievagen B
BDB550524 125 mL Retrievagen A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB550527 Supplier BD Supplier No. 550527
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recommended for zinc-fixed and formalin-fixed paraffin embedded tissue sections that show suboptimal or no reactivity with antibodies for immunohistochemical staining

10% neutral buffered formalin fixes the tissue by cross-linking proteins that may cause a conformational change or “masking” of the antigenic epitope leading to reduced or absent antibody reactivity. The BD Retrievagen Antigen Retrieval Systems are designed to unmask the epitope and cause the enhancement of antibody staining.

  • Retrievagen A (pH 6.0) is provided as two separate solutions that need to be mixed
  • Retrievagen B (pH 9.5) is provided as a ready-to-use solution

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Tris based Buffer with chemical agents for epitope retrieval
Product Type Retrieval Reagent
Quantity 250 mL
For Use With (Application) Immunohistochemical staining
pH 9.5
Regulatory Status RUO
Reagent Type Retrievagen B
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.