Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Retrievagen Antigen Retrieval Systems

Catalog No. p-3901674 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
125 mL
250 mL
Reagent Type:
Retrievagen A
Retrievagen B
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Reagent Type
BDB550524 125 mL Retrievagen A
BDB550527 250 mL Retrievagen B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB550524 Supplier BD Supplier No. 550524
Only null left
Add to Cart
Add to Cart

Recommended for zinc-fixed and formalin-fixed paraffin embedded tissue sections that show suboptimal or no reactivity with antibodies for immunohistochemical staining

10% neutral buffered formalin fixes the tissue by cross-linking proteins that may cause a conformational change or “masking” of the antigenic epitope leading to reduced or absent antibody reactivity. The BD Retrievagen Antigen Retrieval Systems are designed to unmask the epitope and cause the enhancement of antibody staining.

  • Retrievagen A (pH 6.0) is provided as two separate solutions that need to be mixed
  • Retrievagen B (pH 9.5) is provided as a ready-to-use solution

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 25mL Retrievagen A Solution 1 and 100mL Retrievagen A Solution 2
Dilution Take 18 mL of BD Retrievagen A Solution 1 and 82 mL of BD Retrievagen Solution 2 and bring up the volume to 1 L with distilled Water
Product Type Retrieval Reagent
Quantity 125 mL
For Use With (Application) Immunohistochemical staining
pH 6
Regulatory Status RUO
Reagent Type Retrievagen A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.