Learn More
BD ROCK Inhibitor (Y-27632)
Catalog No. BDB562822 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
BDB562822 1 mg
1 options
Catalog No. BDB562822 Supplier BD Supplier No. 562822
Add to Cart
Add to Cart
The ROCK inhibitor, Y-27632, is a selective inhibitor of the p160-Rho-associated coiled kinase (ROCK) and is a factor that enhances embryonic stem cell survival upon single cell dissociation. In addition to increasing cell recovery after dissociation, Y-27632 has been used for various other applications in stem cell research including cryopreservation, sorting, reprogramming, transplantation, and differentiation.
Cell Culture
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Product Type ROCK inhibitor (Y-27632)
Quantity 1 mg