Learn More
BD Saf-T-Intima™ IV Catheter Systems
Catalog No. B383323 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Needle Gauge:
20 G
22 G
Color:
Blue
Pink
Length (English):
0.75 in.
1 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color Length (English)
B383323 22 G Blue 0.75 in.
B383336 20 G Pink 1 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B383323 Supplier BD Supplier No. 383323
Add to Cart
Add to Cart
This item is not returnable. View return policy
With Y Adapter
  • Combines needlestick safety with a closed system design
  • Results in a virtually bloodless procedure for vascular access
  • Made of clinically provenBD Vialon™ biomaterial
TRUSTED_SUSTAINABILITY
Specifications
Material Vialon Biomaterial
Color Blue
Needle Gauge 22 G
For Use With (Application) Virtually bloodless procedure for vascular access, reduces the risks of infiltration, reduces IV restarts due to reduced complications
Includes Flexible textured wings, needle shield, extension tubing and Y adapter
Latex Free Yes
Length (English) 0.75 in.
Product Type IV Catheter System
Disposable Single-use
Quantity 25/Pk.
Needle Point Style Notched
Connection Type Catheter
Sterility Sterile