Learn More
BD Sputum Collection System
Catalog No. 1443220 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Max. RCF:
3500 rpm
Catalog No. Max. RCF
14-432-20 3500 rpm
1 options
This item is not returnable. View return policy
Catalog No. 14-432-20 Supplier BD Supplier No. 290020
This item is not returnable. View return policy
For collecting, transporting and processing biological specimens
  • Unit consists of graduated 50mL screw-cap tube, funnel with hinged lid, label and protective body packaged in protective bag to reduce possible contamination
  • Specimen remains securely contained within the collection tube during transport to the laboratory and is delivered already in the processing tube ready for digestion and centrifugation
  • Tube can be centrifuged to 3500rpm
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sputum Collection System
Closure Type Screw Cap
For Use With (Application) For collecting, transporting and processing biological specimens
Includes Graduated 50 mL screw cap tube, funnel with hinged lid, label and protective body packaged in protective bag
Max. RCF 3500 rpm