Learn More
BD Cell Analysis 3P BD Stem Cell Enumeration Kit, Reliable CD34+ Cell Enumeration, Compatible with BD Flow Cytometers, 50 Tests
Catalog No. NC1252983
Encompass
The BD Stem Cell Enumeration Kit is an IVD solution that accurately enumerates viable dual-positive CD45+/CD34+ hematopoietic stem cell populations using flow cytometry. It provides reliable and standardized results, aiding quality assessment in stem cell transplantation processes. The kit minimizes errors through automated calculations and supports analysis of multiple specimen types, differentiating it with its compatibility with various BD flow cytometers and streamlined workflow following ISHAGE guidelines.
Catalog No. NC1252983 Supplier BD Cell Analysis 3P Supplier No. 344563
Add to Cart
Add to Cart
The BD Stem Cell Enumeration Kit is an IVD solution that accurately enumerates viable dual-positive CD45+/CD34+ hematopoietic stem cell populations using flow cytometry. It provides reliable and standardized results, aiding quality assessment in stem cell transplantation processes. The kit minimizes errors through automated calculations and supports analysis of multiple specimen types, differentiating it with its compatibility with various BD flow cytometers and streamlined workflow following ISHAGE guidelines.