Learn More
BD Stemflow™ Human Pluripotent Stem Cell Sorting and Analysis Kit
Shop All Waters Corporation ProductsDescription
Includes
FITC SSEA-1, PE SSEA-3, Alexa Fluor™ 647 TRA-1-81, FITC isotype control, PE isotype control, Alexa Fluor 647 isotype control, Anti-mouse beads, Anti-rat beads, Negative beads
This kit provides reagents necessary to perform multicolor flow cytometry on human pluripotent stem cells such as human embryonic stem cells (hESCs) and induced pluripotent stem (iPS) cells.
- Reactivity: human
- Size: 50 tests
Specifications
Specifications
| Includes | FITC SSEA-1, PE SSEA-3, Alexa Fluor™ 647 TRA-1-81, FITC isotype control, PE isotype control, Alexa Fluor™ 647 isotype control, Anti-mouse beads, Anti-Rat beads, negative beads |
| Quantity | 50 Tests |