Learn More
Cole-Parmer BD Biocoat Disposable Syringe, Non-Sterile, Luer-Lok, 20 mL, 25/Pk
Catalog No. NC1735865 Shop All Cole-Parmer Products
Encompass_Preferred
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-Lok or slip-tip versions. Syringes come nonsterilized in bulk packages and can be autoclaved. Needles and shield tips are not included; order separately.
This item is not returnable. View return policy
Catalog No. NC1735865 Supplier Cole-Parmer Supplier No. 0794416
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-Lok or slip-tip versions. Syringes come nonsterilized in bulk packages and can be autoclaved. Needles and shield tips are not included; order separately.