Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD T Cell Kit Lyophilized Cells

Catalog No. BDB560760 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560760 10 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560760 Supplier BD Supplier No. 560760
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

5 vials human treated and untreated control cells

Intended to serve as biological controls for antibody staining in BD Phosflow studies designed to analyze cell signaling events associated with T-cell activation

Intracellular Staining, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Quantity 10 Each
Regulatory Status RUO
Content And Storage Store unopened vials of lyophilized cells at 2 - 8°C. Do not freeze.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.